Learn More
Chlormethiazole hydrochloride, Tocris Bioscience™
Potentiates GABAA receptor function
Brand:  Tocris (Bio-Techne) 088110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13448103
  • £88.20 / 10mg
Estimated Shipment date
from Supplier 20-11-2019
Description
Description
Sedative and anticonvulsant which is neuroprotective in a number of animal models. Prevents the degeneration of serotonergic nerve terminals induced by MDMA ('Ecstasy').
Specifications
Chemical Identifiers
6001-74-7
198.105
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
C6H9Cl2NS
OFXYKSLKNMTBHK-UHFFFAOYSA-N
19035073
CC1=C(SC=N1)CCCl.Cl
Specifications
10mg
6001-74-7
chlormethiazole hydrochloride, clomethiazole hydrochloride, somnevrin hydrochloride, 5-2-chloroethyl-4-methylthiazole hydrochloride, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride, 2-4-methylthiazol-5-yl ethyl chloride hydrochloride, clomethiazole hydrochloride mi, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride, thiazole, 5-2-chloroethyl-4-methyl-, hydrochloride 1:1, 5-2-chloroethyl-4-methyl-1,3-thiazole hydrochloride 1:1
CC1=C(SC=N1)CCCl.Cl
198.105
198.11
Chlormethiazole hydrochloride
C6H9Cl2NS
OFXYKSLKNMTBHK-UHFFFAOYSA-N
5-(2-chloroethyl)-4-methyl-1,3-thiazole;hydrochloride
19035073
>99%