missing translation for 'onlineSavingsMsg'
Learn More

Chlorisondamine diiodide, Tocris Bioscience™

Nicotinic antagonist; slow offset

Brand:  Tocris (Bio-Techne) 100150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428263

  • £396.90 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

An exceptionally long lasting nicotinic antagonist (IC50 ∽ 1.6 mM); blockade of central nicotinic responses induced by chlorisondamine can persist for several weeks.
Specifications

Chemical Identifiers

96750-66-2
611.937
chlorisondamine diiodide, 4,5,6,7-tetrachloro-1,3-dihydro-2-methyl-2-2-trimethylammonium ethyl-2h-isoindolium diiodide, chlorisondamine diiodide hplc , white solid, 3-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine diiodide, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-2-trimethylammonio ethyl-2h-isoindolium diiodide, 4,5,6,7-tetrachloro-2-methyl-2-2-trimethylazaniumyl ethyl-2,3-dihydro-1h-isoindol-2-ium diiodide
trimethyl-[2-(4,5,6,7-tetrachloro-2-methyl-1,3-dihydroisoindol-2-ium-2-yl)ethyl]azanium;diiodide
C14H20Cl4I2N2
FPNVAOZHQUJJJQ-UHFFFAOYSA-L
16078959
C[N+]1(CC2=C(C1)C(=C(C(=C2Cl)Cl)Cl)Cl)CC[N+](C)(C)C.[I-].[I-]

Specifications

50mg
96750-66-2
chlorisondamine diiodide, 4,5,6,7-tetrachloro-1,3-dihydro-2-methyl-2-2-trimethylammonium ethyl-2h-isoindolium diiodide, chlorisondamine diiodide hplc , white solid, 3-4-chlorophenyl piperazin-1-yl methyl-1h-pyrrolo 2,3-b pyridine diiodide, 4,5,6,7-tetrachloro-2,3-dihydro-2-methyl-2-2-trimethylammonio ethyl-2h-isoindolium diiodide, 4,5,6,7-tetrachloro-2-methyl-2-2-trimethylazaniumyl ethyl-2,3-dihydro-1h-isoindol-2-ium diiodide
C[N+]1(CC2=C(C1)C(=C(C(=C2Cl)Cl)Cl)Cl)CC[N+](C)(C)C.[I-].[I-]
611.937
611.95
Chlorisondamine diiodide
C14H20Cl4I2N2
FPNVAOZHQUJJJQ-UHFFFAOYSA-L
trimethyl-[2-(4,5,6,7-tetrachloro-2-methyl-1,3-dihydroisoindol-2-ium-2-yl)ethyl]azanium;diiodide
16078959
>99%
Videos
Documents