missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Chlorine in Heavy Mineral Oil standard solution, Specpure™, 50,000μg/g (5.00%)

Brand:  Alfa Aesar™ 042185.AE

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15423875

  • £115.30 / 100mL


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Specifications

Chlorine in Heavy Mineral Oil standard solution
Specpure™
100mL
50,000μg/g (5.00%)
MFCD02264398
Videos
SDS