missing translation for 'onlineSavingsMsg'
Learn More

Chloramphenicol, SPEX CertiPrep™

Catalog Number 15296266
Click to view available options
Quantity:
1 mL
This item is not returnable. View return policy
This item is not returnable. View return policy
Type Chloramphenicol
Physical Form Liquid
Odor Characteristic
Potency 1000μg/mL
Concentration 1000 μg/mL
Quantity 1 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • CHLORAMPHENICOL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : CHLORAMPHENICOL, methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'