missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloramine-T trihydrate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
Specifications
Type | Chloramine T Trihydrate |
Physical Form | Crystalline Powder |
pH | 8 to 10.3 (5% soln.) |
Melting Point | 170.0°C to 177.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97+% |
Linear Formula | CH3C6H4SO2N(Cl)Na·3H2O |
Merck Index | 15, 2075 |
Packaging | Plastic Bottle |
IUPAC Name | N-chloro-4-methylbenzenesulfonamide;sodium;trihydrate |
Show More |
Safety and Handling
Product Identifier
- Chloramine-T trihydrate
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
- EUH031-Contact with acids liberates toxic gas.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'