Learn More
Chloramine-T, Sodium Salt, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Brand: Fisher Chemical C/4320/53
Code : 21
Additional Details : CAS Number : 127-65-1 Weight : 0.50000kg Transport : UN number : 3263 Chem class : 8 Pack group : III
Specifications
7080-50-4 | |
149066 | |
Powder Solid | |
White | |
8 to 10 |
C7H7ClNNaO2S·3H2O | |
281.69g/mol | |
Plastic powder jar | |
170°C | |
500g |