missing translation for 'onlineSavingsMsg'
Learn More

Chitosan, molecular weight: 600,000-800,000, ACROS Organics™

Brand:  Acros Organics 428855000

Code : 29

Additional Details : CAS Number : 9012-76-4 Weight : 0.55000kg

Quantity 500g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10616290

  • £218.60 / 500g


Estimated Shipment
01-02-2018
Add to basket

Specifications

Chemical Identifiers

Chitosan
(C6H11NO4)n
Poly(beta-(1, 4)-2-amino-2-deoxy-D-glucose)
≥90%
MFCD00161512

Specifications

Chitosan
1.5% max.
≥90%
MFCD00161512
Authentic
Plastic bottle
Off-white
9012-76-4
Powder
(C6H11NO4)n
Poly(beta-(1, 4)-2-amino-2-deoxy-D-glucose)
10.0% max.
50 to 200mPa&middnot;s
500g
Videos
SDS