missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Chitinase 3-like 1/YKL-40 Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Chitinase 3-like 1 |
---|---|
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Chitinase 3-like 1 Polyclonal specifically detects Chitinase 3-like 1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Chitinase 3-like 1 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Q61362 | |
1116 | |
Mouse myeloma cell line NS0-derived recombinant mouse Chitinase 3-like 1 Tyr22-Ala381 Accession # Q61362 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Chitinase 3-like 1 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human Chitinase 3-like 1 is observed. |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Sheep | |
Mouse | |
AW208766, Brp39, Chi3l1, Chitinase 3 like 1, gp39, HCgp39, YKL-40 | |
Chil1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only