missing translation for 'onlineSavingsMsg'
Learn More

Chikungunya Virus E1 Antibody (988133) [DyLight 488], Novus Biologicals™

Product Code. 30764575 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30764575 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30764575 Supplier Novus Biologicals Supplier No. FAB97791K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chikungunya Virus E1 Monoclonal antibody specifically detects Chikungunya Virus E1 in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chikungunya Virus E1
Applications ELISA
Classification Monoclonal
Clone 988133
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Chikungunya Virus
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived transfected with viral Chikungunya Virus E1, Tyr810-Gly1224, Accession # Q8JUX5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.