Learn More
Chicken anti-Mouse IgG Fc Secondary Antibody, Biotin, Invitrogen™
Chicken Polyclonal Secondary Antibody
Brand: Invitrogen SA172028
Code : LD
Additional Details : Weight : 0.01000kg
Description
SA1-72028 detects Mouse IgG Fc fragment in mouse samples. SA1-72028 has been successfully used in ELISA and western blot procedures. The SA1-72028 immunogen is from purified mouse IgG isolated from normal mouse IgG.
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Mouse IgG Fc | |
Polyclonal | |
Biotin | |
Purified mouse IgG isolated from normal mouse IgG. | |
500 μg | |
Secondary | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgY |
ELISA, Western Blot | |
1 mg/mL | |
Chicken | |
Purified | |
RUO | |
Mouse | |
Liquid |