missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSA, Chicken, Polyclonal Antibody, Abnova™
Chicken polyclonal antibody raised against native BSA.
Brand: Abnova PAB29123.500ug
Additional Details : Weight : 0.02000kg
Specifications
BSA | |
Polyclonal | |
Chicken polyclonal antibody raised against native BSA. | |
In PBS, pH 7.2 (0.075% sodium azide). | |
Native purified bovine serum albumin with Freund's adjuvant. | |
500 μg | |
Primary | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
ELISA (1:200-1:500 for capture; 1:1000-1:20000 for primary antibody) The optimal working dilution should be determined by the end user. | |
Chicken | |
Immunoaffinity Purification | |
RUO | |
Bovine | |
Liquid |
For Research Use Only