missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ Chemistry Pumping Unit, PC 511 Select

An economical, compact solution for simultaneously running two independent vacuum applications with a single pump. One port is electronically controlled, the second is manually controlled. Includes an exhaust vapor condenser for solvent recovery.

Brand:  VACUUBRAND™ 20733252


Additional Details : Weight : 18.00000kg

 View more versions of this product

Product Code. 16251772

  • £5365.00 / Each
Estimated Shipment: 30-07-2024
to see stock.
Add to basket

Description

Description

  • VACUU·SELECT Vacuum Controller with graphical user interface simplifies work in the laboratory
  • Predefined applications save time and give reproducible results
  • Create unique process sequences with an easy-to-use application editor
  • Powerful vacuum pump with excellent chemical and condensate compatibility
  • For solvent evaporation, the electronically controlled port detects the boiling pressure and switches to two point control to hold pressure steady
  • Chemistry pumping units use chemical resistant solenoid valves to maintain process control
  • Pressure is maintained at the level where the first boiling point is detected by the controller
  • Glass inlet separator with a robust protective coating prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Exhaust vapor condenser allows for near complete recovery of solvents when used with a chilled water line or recirculating chiller
  • Use manual set-point-control for simple processes
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
Specifications

Specifications

7 Mbar (Without Gas Ballast), 12 Mbar (With Gas Ballast)
21 kg
1500 to 1800 min-1
435 x 243 x 457 mm
PC 511 Select
UK
18.2 kg
10°C
50-60 Hz
IP40
2
230 V
VACUU·SELECT
Ready-to-Use Pumping Unit (Completely Mounted with Manual)
CE-certification; II 3/-G IIC T3 x Internal Atm. Only
PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
45 dBA
0.18 kW
40°C
2 m3/hr
Diaphragm Pump
2
1.1 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers