Cheminert™ Model C2 Microbore Injectors
Use Thermo Scientific™ Cheminert™ Model C2 Microbore Injectors as injectors or switching valves.
Overview
Brand: Thermo Scientific
Manufacturer Part Number: C2-1000
VALCO 10 PORT MICROBOREINJECTOR MODEL C2 1/16'' FITTIN.
UNSPSC: 41115700
Code: 16
Additional Details:
Additional Details: Weight: 0.50000kg
Disclaimers: Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Code. 10417534
Quantity Price
1 £ 830.0 / Each
Estimated Shipment
31-03-2017
Add to basket
Description and Specification
Specification
Item Description Model C2 Injector
Volume (Metric) Sample 5µL Loop
Actuation Manual
Ports 10
Stator Material N60 stainless
  • 1/16″ fittings
  • 0.010″ ports
  • Available in 6-port or 10-port configurations
  • Available with manual or microelectric actuator