missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Chemicon™ TMB/E Single Reagent Horseradish Peroxidase Substrate
Blue colored TMB/E Single Reagent Horseradish Peroxidase Substrate (soluble) for ELISA
Brand: Merck Millipore ES001-500ML
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- ES001 is a single component reagent stable at room temperature and not sensitive to normal laboratory light.
- It is optimized with respect to TMB and hydrogen peroxide concentrations and yields a linear response with the concentrations of HRP usually employed in immunological assays.
- λmax 450 nm
- Color Code: Yellow