Learn More
CHEMetrics CHEMets Ammonia Test Kit Refill
Provides results in two minutes or less
Brand: CHEMetrics R1501
Additional Details : CAS Number : 1310-73-2 Weight : 0.01500kg Transport : UN number : 1760 Chem class : 8 Pack group : II
Alert:
Contains mercury. Dispose according to local, state or federal laws.
Description
This product contains 30 self-filling CHEMets ampules to refill kit K-1510 for the analysis of ammonia. R-1501 is compatible with C-1501 and C-1510 comparators.Specifications
Ammonia | |
30 self-filling CHEMets ampoules to refill kit K-1510 | |
0 to 1 ppm and 1 to 10 ppm | |
1 min. |
C-1501 and C-1510 comparators | |
30 | |
Direct nesslerization | |
CHEMets Refill Kit |
Safety and Handling
- ammonia CHEMets & VACUettes refills and ammonia vacu-vials ampoules
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, sodium borate, dehydrate, mercuric iodide,potassium iodide
Warning! Prop 65