Learn More
CHCHD10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA551935
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GLMAQMATTA AGVAVGSAVG HVMGSALTGA FSGGSSEPSQ PAVQQAPTPA APQPLQMGPC AYEIRQFLDC STTQSDLSLC EGFSEALKQC KYYHGLSSLP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
CHCHD10 Polyclonal antibody specifically detects CHCHD10 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CHCHD10 | |
Polyclonal | |
Unconjugated | |
CHCHD10 | |
1620401E04Rik; AI267078; C22orf16; CHCHD10; coiled-coil-helix-coiled-coil-helix domain containing 10; coiled-coil-helix-coiled-coil-helix domain-containing protein 10, mitochondrial; FTDALS2; IMMD; N27C7-4; Ndg2; Nur77 downstream gene 2; Nur77 downstream protein 2; Protein N27C7-4; SMAJ | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
400916 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WYQ3 | |
CHCHD10 | |
Recombinant protein corresponding to Human CHCHD10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only