missing translation for 'onlineSavingsMsg'
Learn More

CGS 19755, Tocris Bioscience™

Potent competitive NMDA antagonist

Brand:  Tocris (Bio-Techne) 124150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13438953

  • £646.37 / 50mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, competitive NMDA receptor antagonist. Anticonvulsant and neuroprotective. Also available as part of the Mixed NMDA Receptor Tocriset™ .
Specifications

Chemical Identifiers

110347-85-8
223.165
selfotel, selfotel usan:inn, 2s,4r-4-phosphonomethyl piperidine-2-carboxylic acid, cis-4-phosphonomethyl pipecolic acid, chembl39664, 2-piperidinecarboxylic acid, 4-phosphonomethyl-, cis, 2-piperidinecarboxylic acid, 4-phosphonomethyl-, 2s,4r, cis-4-phosphonomethyl piperidine-2-carboxylic acid, cis-4-phosphonomethyl-piperidine-2-carboxylic acid, selfotel, +
CHEBI:34973
C1CNC(CC1CP(=O)(O)O)C(=O)O
C7H14NO5P
LPMRCCNDNGONCD-RITPCOANSA-N
68736
(2S,4R)-4-(phosphonomethyl)piperidine-2-carboxylic acid

Specifications

CGS 19755
110347-85-8
selfotel, selfotel usan:inn, 2s,4r-4-phosphonomethyl piperidine-2-carboxylic acid, cis-4-phosphonomethyl pipecolic acid, chembl39664, 2-piperidinecarboxylic acid, 4-phosphonomethyl-, cis, 2-piperidinecarboxylic acid, 4-phosphonomethyl-, 2s,4r, cis-4-phosphonomethyl piperidine-2-carboxylic acid, cis-4-phosphonomethyl-piperidine-2-carboxylic acid, selfotel, +
C1CNC(CC1CP(=O)(O)O)C(=O)O
223.165
CHEBI:34973
>98%
50mg
C7H14NO5P
LPMRCCNDNGONCD-RITPCOANSA-N
(2S,4R)-4-(phosphonomethyl)piperidine-2-carboxylic acid
68736
223.17
Videos