Learn More
CGP 54626 hydrochloride, Tocris Bioscience™
Potent, selective GABAB antagonist
Brand:  Tocris (Bio-Techne) 1088/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13488533
  • £183.48 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
A potent, selective GABAB receptor antagonist (IC50 = 4 nM).
Specifications
Chemical Identifiers
149184-21-4
444.758
[S-(R™, R™)]-[3-[[1-(3, 4-Dichlorophenyl)ethyl]amino]-2-hydroxypropyl](cyclohexylmethyl) phosphinic acid
C18H29Cl3NO3P
ZQCFHOVIXCJPLE-LINSIKMZSA-N
197583
Specifications
CGP 54626 hydrochloride
149184-21-4
C18H29Cl3NO3P
ZQCFHOVIXCJPLE-LINSIKMZSA-N
cyclohexylmethyl-[(2S)-3-[[(1S)-1-(3,4-dichlorophenyl)ethyl]amino]-2-hydroxypropyl]phosphinic acid;hydrochloride
197583
10mg
>96%
[S-(R™, R™)]-[3-[[1-(3, 4-Dichlorophenyl)ethyl]amino]-2-hydroxypropyl](cyclohexylmethyl) phosphinic acid
CC(C1=CC(=C(C=C1)Cl)Cl)NCC(CP(=O)(CC2CCCCC2)O)O.Cl
444.758
444.76
Videos