missing translation for 'onlineSavingsMsg'
Learn More

CGP 54626 hydrochloride, Tocris Bioscience™

Potent, selective GABAB antagonist

Brand:  Tocris (Bio-Techne) 1088/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13488453

  • £671.97 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

A potent, selective GABAB receptor antagonist (IC50 = 4 nM).
Specifications

Chemical Identifiers

149184-21-4
444.758
[S-(R™, R™)]-[3-[[1-(3, 4-Dichlorophenyl)ethyl]amino]-2-hydroxypropyl](cyclohexylmethyl) phosphinic acid
cyclohexylmethyl-[(2S)-3-[[(1S)-1-(3,4-dichlorophenyl)ethyl]amino]-2-hydroxypropyl]phosphinic acid;hydrochloride
C18H29Cl3NO3P
ZQCFHOVIXCJPLE-LINSIKMZSA-N
197583
CC(C1=CC(=C(C=C1)Cl)Cl)NCC(CP(=O)(CC2CCCCC2)O)O.Cl

Specifications

CGP 54626 hydrochloride
149184-21-4
C18H29Cl3NO3P
ZQCFHOVIXCJPLE-LINSIKMZSA-N
cyclohexylmethyl-[(2S)-3-[[(1S)-1-(3,4-dichlorophenyl)ethyl]amino]-2-hydroxypropyl]phosphinic acid;hydrochloride
197583
50mg
>96%
[S-(R™, R™)]-[3-[[1-(3, 4-Dichlorophenyl)ethyl]amino]-2-hydroxypropyl](cyclohexylmethyl) phosphinic acid
CC(C1=CC(=C(C=C1)Cl)Cl)NCC(CP(=O)(CC2CCCCC2)O)O.Cl
444.758
444.76
Videos