Learn More
CGP 52432, Tocris Bioscience™
Potent, selective GABAB antagonist
Brand:  Tocris (Bio-Techne) 1246/10
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458963
  • £183.48 / 10mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Potent, selective GABAB receptor antagonist (IC50 = 85 nM).
Specifications
Chemical Identifiers
139667-74-6
384.234
tocris-1246, biomol-nt_000239, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorphenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methylamino propyl-diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl
3-[(3,4-dichlorophenyl)methylamino]propyl-(diethoxymethyl)phosphinic acid
C15H24Cl2NO4P
GJZVQXWEIYRHBE-UHFFFAOYSA-N
132252
CCOC(OCC)P(=O)(CCCNCC1=CC(=C(C=C1)Cl)Cl)O
Specifications
CGP 52432
139667-74-6
tocris-1246, biomol-nt_000239, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorphenyl methyl amino propyl diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methylamino propyl-diethoxymethyl phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl phosphinic acid, phosphinic acid, 3-3,4-dichlorophenyl methyl amino propyl diethoxymethyl
CCOC(OCC)P(=O)(CCCNCC1=CC(=C(C=C1)Cl)Cl)O
384.234
384.24
10mg
C15H24Cl2NO4P
GJZVQXWEIYRHBE-UHFFFAOYSA-N
3-[(3,4-dichlorophenyl)methylamino]propyl-(diethoxymethyl)phosphinic acid
132252
>99%
Videos