Learn More
CGP 37849, Tocris Bioscience™
Potent and selective NMDA antagonist
Brand:  Tocris (Bio-Techne) 146910
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13439553
  • £158.76 / 10mg
Estimated Shipment date
from Supplier 28-10-2019
Description
Description
Potent, selective and competitive NMDA receptor antagonist (Ki = 35 nM). Anticonvulsive following oral administration in vivo.
Specifications
Chemical Identifiers
127910-31-0
209.138
4-methyl-appa, e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, 2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono, 3e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, chembl29811, cgp hplc, e-2-amino-4-methyl-5-phosphono-3-pentenoic acid, e-+/--2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, e-+-
(E)-2-amino-4-methyl-5-phosphonopent-3-enoic acid
C6H12NO5P
BDYHNCZIGYIOGJ-DUXPYHPUSA-N
5950212
CC(=CC(C(=O)O)N)CP(=O)(O)O
Specifications
10mg
127910-31-0
4-methyl-appa, e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, 2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono, 3e-2-amino-4-methyl-5-phosphonopent-3-enoic acid, chembl29811, cgp hplc, e-2-amino-4-methyl-5-phosphono-3-pentenoic acid, e-+/--2-amino-4-methyl-5-phosphono-3-pentenoic acid, 3-pentenoic acid, 2-amino-4-methyl-5-phosphono-, e-+-
CC(=CC(C(=O)O)N)CP(=O)(O)O
209.138
209.14
CGP 37849
C6H12NO5P
BDYHNCZIGYIOGJ-DUXPYHPUSA-N
(E)-2-amino-4-methyl-5-phosphonopent-3-enoic acid
5950212
>99%