missing translation for 'onlineSavingsMsg'
Learn More

CGP 37157, Tocris Bioscience™

Antagonist of mitochondrial Na+/Ca2+ exchange

Brand:  Tocris (Bio-Techne) 1114/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458563

  • £194.04 / 10mg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective antagonist of the mitochondrial Na+-Ca2+ exchanger (IC50 = 0.4 μM).
Specifications

Chemical Identifiers

75450-34-9
324.219
7-chloro-5-2-chlorophenyl-1,5-dihydro-4,1-benzothiazepin-2 3h-one, 7-chloro-5-2-chlorophenyl-1,5-dihydro-4,1-benzothiazepin-2-one, 7-chloro-5-2-chlorophenyl-3,5-dihydro-1h-4,1-benzothiazepin-2-one, cgp hplc , powder, 4,1-benzothiazepin-2 3h-one, 7-chloro-5-2-chlorophenyl-1,5-dihydro, 7-chloro-5-2-chlorophenyl-3,5-dihydrobenzo e 1,4 thiazepin-2 1h-one
7-chloro-5-(2-chlorophenyl)-1,5-dihydro-4,1-benzothiazepin-2-one
C15H11Cl2NOS
KQEPIRKXSUIUTH-UHFFFAOYSA-N
2688
C1C(=O)NC2=C(C=C(C=C2)Cl)C(S1)C3=CC=CC=C3Cl

Specifications

10mg
75450-34-9
7-chloro-5-2-chlorophenyl-1,5-dihydro-4,1-benzothiazepin-2 3h-one, 7-chloro-5-2-chlorophenyl-1,5-dihydro-4,1-benzothiazepin-2-one, 7-chloro-5-2-chlorophenyl-3,5-dihydro-1h-4,1-benzothiazepin-2-one, cgp hplc , powder, 4,1-benzothiazepin-2 3h-one, 7-chloro-5-2-chlorophenyl-1,5-dihydro, 7-chloro-5-2-chlorophenyl-3,5-dihydrobenzo e 1,4 thiazepin-2 1h-one
C1C(=O)NC2=C(C=C(C=C2)Cl)C(S1)C3=CC=CC=C3Cl
324.219
324.22
CGP 37157
C15H11Cl2NOS
KQEPIRKXSUIUTH-UHFFFAOYSA-N
7-chloro-5-(2-chlorophenyl)-1,5-dihydro-4,1-benzothiazepin-2-one
2688
>99%
Videos
Documents