missing translation for 'onlineSavingsMsg'
Learn More

CGP 20712 dihydrochloride, Tocris Bioscience™

Highly potent and selective β1 antagonist

Brand:  Tocris (Bio-Techne) 102450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13428323

  • £842.31 / 50mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Highly selective and potent β1-adrenoceptor antagonist (IC50 = 0.7 nM). Displays 10,000-fold selectivity over β2-adrenoceptors. Also available as part of the β-Adrenoceptor Antagonist Tocriset™ .
Specifications

Chemical Identifiers

1216905-73-5
567.387
cgp 20712 dihydrochloride, 1-2-3-carbamoyl-4-hydroxy phenoxy ethylamino-3-4-1-methyl-4-trifluoromethyl-2-imidazolyl phenoxy-2-propanol dihydrochloride
2-hydroxy-5-[2-[[2-hydroxy-3-[4-[1-methyl-4-(trifluoromethyl)imidazol-2-yl]phenoxy]propyl]amino]ethoxy]benzamide;dihydrochloride
C23H27Cl2F3N4O5
PURFQCFKYNMIQF-UHFFFAOYSA-N
56972164
CN1C=C(N=C1C2=CC=C(C=C2)OCC(CNCCOC3=CC(=C(C=C3)O)C(=O)N)O)C(F)(F)F.Cl.Cl

Specifications

50mg
1216905-73-5
cgp 20712 dihydrochloride, 1-2-3-carbamoyl-4-hydroxy phenoxy ethylamino-3-4-1-methyl-4-trifluoromethyl-2-imidazolyl phenoxy-2-propanol dihydrochloride
CN1C=C(N=C1C2=CC=C(C=C2)OCC(CNCCOC3=CC(=C(C=C3)O)C(=O)N)O)C(F)(F)F.Cl.Cl
567.387
567.39
CGP 20712 dihydrochloride
C23H27Cl2F3N4O5
PURFQCFKYNMIQF-UHFFFAOYSA-N
2-hydroxy-5-[2-[[2-hydroxy-3-[4-[1-methyl-4-(trifluoromethyl)imidazol-2-yl]phenoxy]propyl]amino]ethoxy]benzamide;dihydrochloride
56972164
>96%
Videos
Documents