Learn More
CGP 13501, Tocris Bioscience™
Positive modulator at GABAB receptors
Brand:  Tocris (Bio-Techne) 151450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13429593
  • £568.89 / 50mg
Estimated Shipment date
from Supplier 23-03-2020
Description
Description
Positive allosteric modulator of GABAB receptors, potentiating the effects of GABA at this receptor type.
Specifications
Chemical Identifiers
56189-68-5
290.447
3-3,5-ditert-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, tocris-1514, lopac-c-0987, d06vmv, cgp hplc , solid, allosteric gaba b receptor modulators, novartis, 2,2-di-methyl-3-3,5-di-t-butyl-4-hydroxyphenyl propanal, 3-3,5-di-t-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, 3-3,5-di-tert-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, 2,2-dimethyl-3-3,5-di-tert.butyl-4-hydroxyphenyl propionaldehyde
3-(3,5-ditert-butyl-4-hydroxyphenyl)-2,2-dimethylpropanal
C19H30O2
XGWATTXMMMANFJ-UHFFFAOYSA-N
3519541
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(C)(C)C=O
Specifications
50mg
56189-68-5
3-3,5-ditert-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, tocris-1514, lopac-c-0987, d06vmv, cgp hplc , solid, allosteric gaba b receptor modulators, novartis, 2,2-di-methyl-3-3,5-di-t-butyl-4-hydroxyphenyl propanal, 3-3,5-di-t-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, 3-3,5-di-tert-butyl-4-hydroxyphenyl-2,2-dimethylpropanal, 2,2-dimethyl-3-3,5-di-tert.butyl-4-hydroxyphenyl propionaldehyde
CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CC(C)(C)C=O
290.447
290.44
CGP 13501
C19H30O2
XGWATTXMMMANFJ-UHFFFAOYSA-N
3-(3,5-ditert-butyl-4-hydroxyphenyl)-2,2-dimethylpropanal
3519541
>98%