missing translation for 'onlineSavingsMsg'
Learn More

CGP 12177 hydrochloride, Tocris Bioscience™

β3 partial agonist. β1/β2 antagonist.

Brand:  Tocris (Bio-Techne) 1134/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458703

  • £82.35 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Partial agonist at β3-adrenoceptors. β1- and β2-adrenoceptor antagonist (Ki values are 0.9, 4 and 88 nM for β1, β2 and β3 receptors respectively).
Specifications

Chemical Identifiers

64208-32-8
315.798
cgp 12177 hydrochloride, 4-3-tert-butylamino-2-hydroxypropoxy-1h-benzo d imidazol-2 3h-one hydrochloride, +/--cgp-12177a hydrochloride, cgp 12177 hydochloride, y-cgp-12177a hydrochloride, ?-cgp-12177a hydrochloride, inverted exclamation marka-cgp-12177a, 1,3-dihydro-2h-benzimidazol-2-one hydrochloride, inverted question mark-cgp-12177a hydrochloride, +/--cgp-12177a hplc , solid
4-[3-(tert-butylamino)-2-hydroxypropoxy]-1,3-dihydrobenzimidazol-2-one;hydrochloride
C14H22ClN3O3
YQVFCYCTITZLSX-UHFFFAOYSA-N
11957517
CC(C)(C)NCC(COC1=CC=CC2=C1NC(=O)N2)O.Cl

Specifications

CGP 12177 hydrochloride
64208-32-8
cgp 12177 hydrochloride, 4-3-tert-butylamino-2-hydroxypropoxy-1h-benzo d imidazol-2 3h-one hydrochloride, +/--cgp-12177a hydrochloride, cgp 12177 hydochloride, y-cgp-12177a hydrochloride, ?-cgp-12177a hydrochloride, inverted exclamation marka-cgp-12177a, 1,3-dihydro-2h-benzimidazol-2-one hydrochloride, inverted question mark-cgp-12177a hydrochloride, +/--cgp-12177a hplc , solid
CC(C)(C)NCC(COC1=CC=CC2=C1NC(=O)N2)O.Cl
315.798
315.8
10mg
C14H22ClN3O3
YQVFCYCTITZLSX-UHFFFAOYSA-N
4-[3-(tert-butylamino)-2-hydroxypropoxy]-1,3-dihydrobenzimidazol-2-one;hydrochloride
11957517
>98%
Videos