missing translation for 'onlineSavingsMsg'
Learn More

CFM-2, Tocris Bioscience™

Non-competitive AMPA antagonist

Brand:  Tocris (Bio-Techne) 108210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438523

  • £139.05 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Novel, selective non-competitive AMPA antagonist. Highly potent, long-acting anticonvulsant.
Specifications

Chemical Identifiers

178616-26-7
311.341
cfm-2, 1-4-aminophenyl-7,8-dimethoxy-3,5-dihydro-2,3-benzodiazepin-4-one, 1-4'-aminophenyl-3,5-dihydro-7,8-dimethoxy-4h-2,3-benzodiazepin-4-one, cfm 2, tocris-1082, biomol-nt_000179
1-(4-aminophenyl)-7,8-dimethoxy-3,5-dihydro-2,3-benzodiazepin-4-one
C17H17N3O3
MJKADKZSYQWGLL-UHFFFAOYSA-N
4377504
COC1=C(C=C2C(=C1)CC(=O)NN=C2C3=CC=C(C=C3)N)OC

Specifications

CFM-2
178616-26-7
cfm-2, 1-4-aminophenyl-7,8-dimethoxy-3,5-dihydro-2,3-benzodiazepin-4-one, 1-4'-aminophenyl-3,5-dihydro-7,8-dimethoxy-4h-2,3-benzodiazepin-4-one, cfm 2, tocris-1082, biomol-nt_000179
COC1=C(C=C2C(=C1)CC(=O)NN=C2C3=CC=C(C=C3)N)OC
311.341
311.34
10mg
C17H17N3O3
MJKADKZSYQWGLL-UHFFFAOYSA-N
1-(4-aminophenyl)-7,8-dimethoxy-3,5-dihydro-2,3-benzodiazepin-4-one
4377504
>98%
Videos