missing translation for 'onlineSavingsMsg'
Learn More

Cevimeline hydrochloride, Tocris Bioscience™

Selective M1 agonist

Brand:  Tocris (Bio-Techne) 368950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13453424

  • £730.59 / 50mg

Estimated Shipment date
from Supplier 03-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective M1 receptor agonist. Induces atropine-sensitive contractions of isolated guinea pig ilea and trachea preparations (EC50 values are 3.5 and 3 μM respectively). Reverses AF64A-induced cognitive impairments in vivo.
Specifications

Chemical Identifiers

107220-28-0
235.77
hydrogen cevimeline chloride
2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride
C10H18ClNOS
SURWTGAXEIEOGY-UHFFFAOYSA-N
9837584
CC1OC2(CN3CCC2CC3)CS1.Cl

Specifications

50mg
107220-28-0
hydrogen cevimeline chloride
CC1OC2(CN3CCC2CC3)CS1.Cl
235.77
235.77
Cevimeline hydrochloride
C10H18ClNOS
SURWTGAXEIEOGY-UHFFFAOYSA-N
2-methylspiro[1,3-oxathiolane-5,3'-1-azabicyclo[2.2.2]octane];hydrochloride
9837584
>98%
Videos
Documents