missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetyltrimethylammonium bromide, MP Biomedicals™
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
Specifications
Color | White |
Physical Form | Powder |
Assay Percent Range | 0.9906 |
Quantity | 5 g |
Formula Weight | 364.5 |
Chemical Name or Material | Cetyltrimethylammonium bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'