missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetyl Trimethylammonium Bromide, Pure, Fisher Chemical™
180.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
149.09 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
180.04 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
149.09 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
180.04 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
500g, Plastic powder jar
Description
C19H42BrN, CAS Number-57-09-0, 500g, White, 364.45g/mol, 230 deg.C, 11772, Plastic powder jar, 5 to 7, Powder Solid
Specifications
Melting Point | 230°C |
CAS | 57-09-0 |
Color | White |
pH | 5 to 7 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Molecular Formula | C19H42BrN |
MDL Number | MFCD00011772 |
InChI Key | LZZYPRNAOMGNLH-UHFFFAOYSA-M |
SMILES | [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Show More |
Safety and Handling
Product Identifier
- Cetyl trimethylammonium bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'