missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetyl Trimethylammonium Bromide, Certified AR for Analysis, Fisher Chemical™
201.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Specifications
Melting Point | 230°C |
Color | White |
pH | 5 to 7 |
Physical Form | Powder/Solid |
Quantity | 100 g |
Formula Weight | 364.45g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Cetyl trimethylammonium bromide
- Danger
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- H318-Causes serious eye damage.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'