missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Cetrimide Agar
Brand: Thermo Scientific™ PO0551A
Code : 65
Additional Details : Weight : 0.30000kg
Specifications
2°C to 8°C | |
Cetrimide Agar | |
Agar | |
10 Pack |