missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetrelimab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542197
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Cetrelimab (JNJ-63723283) is a Programmed Cell Death Receptor-1 (PD-1) Inhibitor.Specifications
Cetrelimab | |
Recombinant Monoclonal | |
Unconjugated | |
JNJ-63723283 | |
Human PDCD1/PD1/CD279 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG4 κ |