missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium sulfate, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Cesium sulfate is used to prepare dense aqueous solutions, which finds application in isopycnic centrifugation especially for DNA purification and recovery. It acts as a catalyst for sulfur dioxide oxidation with vanadium or vanadium pentoxide. Further, it acts as a metal oxide catalyst enhancer for heterogeneous processes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1010°C |
Color | White |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99.99% (Metal basis) |
Solubility Information | Soluble in water. Insoluble in ethanol and acetone. |
IUPAC Name | dicesium;sulfate |
Formula Weight | 361.87 |
Sensitivity | Hygroscopic |
Appearance | White powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'