missing translation for 'onlineSavingsMsg'
Learn More

Cesium silicate (meta), 99%, Thermo Scientific Chemicals

Catalog Number p-7050990
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
15586-77-3
Cs2O3Si
341.89
MFCD00798519
MZRJYMUQLBFTMG-UHFFFAOYSA-N
dicaesium oxosilanebis olate, caesium silicate, dicaesium 1+ ion oxosilanebis olate, cesium metasilicate trace metals basis 100g
12893658
dicesium;dioxido(oxo)silane
[Cs+].[Cs+].[O-][Si]([O-])=O
This item is not returnable. View return policy
15586-77-3
Cs2O3Si
341.89
MFCD00798519
MZRJYMUQLBFTMG-UHFFFAOYSA-N
dicaesium oxosilanebis olate, caesium silicate, dicaesium 1+ ion oxosilanebis olate, cesium metasilicate trace metals basis 100g
12893658
dicesium;dioxido(oxo)silane
[Cs+].[Cs+].[O-][Si]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 341.89
Percent Purity 99%
Odor Odorless
Chemical Name or Material Cesium silicate (meta)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'