missing translation for 'onlineSavingsMsg'
Learn More

Cesium, plasma standard solution, Cs 1000μg/mL, Thermo Scientific Chemicals, Specpure™

Catalog Number p-4824669
Click to view available options
Quantity:
50 mL
500 mL
This item is not returnable. View return policy
534-17-8, 7697-37-2, 7732-18-5
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
[Cs+].[Cs+].[O-]C([O-])=O
This item is not returnable. View return policy
534-17-8, 7697-37-2, 7732-18-5
CCs2O3
325.82
MFCD00010957
FJDQFPXHSGXQBY-UHFFFAOYSA-L
10796
[Cs+].[Cs+].[O-]C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
Color Colorless
IUPAC Name dicesium;carbonate
Concentration or Composition (by Analyte or Components) Nitric acid: 5.0%; Cesium carbonate: 0.122%; Water: 94.878%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Physical Form Liquid
Chemical Name or Material Cesium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'