missing translation for 'onlineSavingsMsg'
Learn More

Cesium, plasma standard solution, Cs 1000μg/mL, Alfa Aesar™, Specpure™

Brand:  Alfa Aesar 013825.AE

Code : NEW

Additional Details : Weight : 0.01000kg Transport : UN number : 3264 Chem class : 8 Pack group : III

Quantity 100mL

 View more versions of this product

Product Code. 11381369

  • £50.90 / 100mL


Estimated Shipment date
from Supplier 19-07-2018
Add to basket

Specifications

Chemical Identifiers

Cesium
Liquid
MFCD00134037
7732-18-5
Cs2CO3 in 5% HNO3

Specifications

Cesium
Clear liquid
Cs2CO3 in 5% HNO3
Nitric acid: 5.0%; Cesium carbonate: 0.122%; Water: 94.878%
Clear colorless
7732-18-5
Liquid
MFCD00134037
23 hPa (17mm Hg) at 20°C
100mL
Videos
SDS