missing translation for 'onlineSavingsMsg'
Learn More

Cesium perchlorate 99% , ACROS Organics™

Brand:  Acros Organics 389760100

Code : 29

Additional Details : CAS Number : 13454-84-7 Weight : 0.06000kg Transport : UN number : 1481 Chem class : 1 Pack group : II


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10463555

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Cesium perchlorate
99%
CsClO4
MFCD00010956
99%
ClCsO4
232.36

Specifications

Cesium perchlorate
Crystals or Crystalline Powder
99%
CsClO4
MFCD00010956
Solubility in water: 1.9g/100mL.
250.0°C
13454-84-7
99%
ClCsO4
232.36
Glass bottle
White
10g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Cesium perchlorate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS