Cesium perchlorate 99% , ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 389760500

50GR Cesium perchlorate, 99%

UNSPSC: 12161500

Code: 29

Additional Details:
Additional Details: CAS Number: 13454-84-7 Weight: 0.10000kg Transport: UN number : 1481 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 50g

Product Code. 10310244

Quantity Price
1 £ 67.9 / 50g
Estimated Shipment date
from Supplier 03-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Cesium perchlorate
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula ClCsO4
Linear Formula CsClO4
Formula Weight 232.36
MDL Number MFCD00010956

Specification

Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Formula Weight 232.36
Quantity 50g
Color White
Linear Formula CsClO4
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 13454-84-7
Assay Percent Range 99%
MDL Number MFCD00010956
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
Molecular Formula ClCsO4
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Cesium perchlorate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.