missing translation for 'onlineSavingsMsg'
Learn More

Cesium hydroxide, 50% w/w aq. soln., 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048279
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
21351-79-1
CsHO
149.91
MFCD00010964
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
62750
CHEBI:33988
cesium;hydroxide
[OH-].[Cs+]
This item is not returnable. View return policy
21351-79-1
CsHO
149.91
MFCD00010964
HUCVOHYBFXVBRW-UHFFFAOYSA-M
cesium hydroxide, caesium hydroxide, cesium hydrate, csoh, cesium hydroxide cs oh, cesium hydroxide dimer, cesium hydoxide, cs.ho, caesium 1+ oh, cesium hydroxide solution
62750
CHEBI:33988
cesium;hydroxide
[OH-].[Cs+]

Cesium hydroxide is used in electric storage batteries. Also used to make porous cesium titanosilicates and potential ion-exchange materials for cleanup of radioactive wastes. Cesium Hydroxide is used as electrolyte in alkaline batteries at subzero temperatures, color photography and as a catalyst in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Quantity 25 g
UN Number UN2681
Solubility Information Highly soluble in water.
Formula Weight 149.91
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Sensitivity Air Sensitive
Density 1.725 g/mL
Assay (metals basis)
Chemical Name or Material Cesium hydroxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cesium hydroxide, 50% w/w Aqueous Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Cesium hydroxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'