Cesium formate monohydrate, for analysis, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 392800250

25GR Cesium formate monohydrate, for analysis

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: Weight: 0.07500kg


Packaging Plastic bottle
Quantity 25g

Product Code. 10589593

Quantity Price
1 £ 23.8 / 25g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Cesium formate monohydrate
Purity Grade For analysis
Assay ≥99.5%
Molecular Formula CHCsO2·H2O
Linear Formula HCOOCs
Formula Weight 195.94
MDL Number MFCD00039103

Specification

Assay ≥99.5%
Physical Form Crystalline Powder
Formula Weight 195.94
Quantity 25g
Color White
Sodium (Na) 0.02% max.
Barium (Ba) 0.005% max.
Chloride 0.03% max.
Rubidium (Rb) 0.005% max.
Linear Formula HCOOCs
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Magnesium (Mg) 0.0002% max.
Purity Grade For analysis
Strontium (Sr) 0.0005% max.
Calcium (Ca) 0.001% max.
Potassium (K) 0.03% max.
MDL Number MFCD00039103
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Lithium (Li) 0.001% max.
Molecular Formula CHCsO2·H2O
Chemical Name or Material Cesium formate monohydrate
Sulfate 0.01% max.
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.