missing translation for 'onlineSavingsMsg'
Learn More

Cesium formate hydrate, White chunks/powder, 99.9% (Metals basis), Alfa Aesar™

Brand:  Alfa Aesar 012927.22

Code : NEW

Additional Details : CAS Number : 3495-36-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11382008

  • £95.80 / 100g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

Cesium formate hydrate
99.9% (Metals basis)
177.92
Chunks and Powder
CsOOCH·xH2O
MFCD00039103

Specifications

Cesium formate hydrate
99.9% (Metals basis)
177.92
Hydrate
100g
Chunks and Powder
CsOOCH·xH2O
MFCD00039103
White
Videos
SDS