missing translation for 'onlineSavingsMsg'
Learn More

Cesium fluoride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7048931
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
13400-13-0
CsF
151.90
MFCD00010960
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
25953
cesium;fluoride
[F-].[Cs+]
This item is not returnable. View return policy
13400-13-0
CsF
151.90
MFCD00010960
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
25953
cesium;fluoride
[F-].[Cs+]

Cesium fluoride is used for desilylation reactions in organic synthesis. Useful base in organic chemistry, due to its low nucleophilicity. Owing to its high ionic character, it is a very reactive source of fluoride ion in organic synthesis. Reagent of choice when reactive naked fluoride is required. Effective catalyst in promoting Knoevenegal condensation. Single crystals of CsF are useful as the windows of cells for infrared spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1251°C
Melting Point 682°C
Color White
pH 6.5 to 7.5
Physical Form Crystalline
Quantity 1 g
UN Number UN2923
Solubility Information Soluble in water. Insoluble in dioxane and pyridine.
Formula Weight 151.9
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium fluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'