missing translation for 'onlineSavingsMsg'
Learn More

Cesium fluoride, 99%, for analysis, Thermo Scientific Chemicals

Catalog Number p-4509657
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13400-13-0
CsF
151.90
MFCD00010960
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
25953
[F-].[Cs+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
13400-13-0
CsF
151.90
MFCD00010960
XJHCXCQVJFPJIK-UHFFFAOYSA-M
cesium fluoride, caesium fluoride, cesium monofluoride, dicesium difluoride, tricesium trifluoride, unii-t76a371hjr, caesium 1+ ion fluoride, caesiumfluoride, cesiumfluoride
25953
[F-].[Cs+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Boiling Point 1251.0°C
Melting Point 682.0°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98.5% min
Linear Formula CsF
Fieser 01,121; 06,100; 07,57; 08,81; 09,100; 10,81; 11,115; 12,108; 13,68; 14,79; 15,75; 16,69; 17,68
Merck Index 15,2011
Solubility Information Solubility in water: 369g/100mL (18°C). Other solubilities: soluble in methanol, insoluble in dioxane and pyridine
IUPAC Name caesium(1+) fluoride
Formula Weight 151.91
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Cesium fluoride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'