missing translation for 'onlineSavingsMsg'
Learn More

Cesium dichromate, 99.8% (metals basis), Thermo Scientific™

Catalog Number p-7051002
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
13530-67-1
Cr2Cs2O7
481.80
MFCD00049436
AQWIYQPYXQGBQU-UHFFFAOYSA-N
cesium dichromate, caesium dichromate, acmc-20alhd, cr2o7.2cs, dicaesium 1+ chromate, chromic acid h2cr2o7 ,cesium salt 1:2, dicesium oxido-oxido dioxo chromio oxy-dioxochromium
3084158
dicesium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[Cs+].[Cs+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
This item is not returnable. View return policy
13530-67-1
Cr2Cs2O7
481.80
MFCD00049436
AQWIYQPYXQGBQU-UHFFFAOYSA-N
cesium dichromate, caesium dichromate, acmc-20alhd, cr2o7.2cs, dicaesium 1+ chromate, chromic acid h2cr2o7 ,cesium salt 1:2, dicesium oxido-oxido dioxo chromio oxy-dioxochromium
3084158
dicesium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
[Cs+].[Cs+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Coarse Powder
Quantity 25 g
Linear Formula Cs2Cr2O7
UN Number UN3087
Formula Weight 481.8
Percent Purity 99.8%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cesium dichromate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'