missing translation for 'onlineSavingsMsg'
Learn More

Cesium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050133
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]
This item is not returnable. View return policy
7647-17-8
ClCs
168.36
MFCD00010955
AIYUHDOJVYHVIT-UHFFFAOYSA-M
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride
24293
CHEBI:63039
cesium;chloride
[Cl-].[Cs+]

It is used as a precursor for cesium metal, it act as a phase transfer catalyst, for molecular genetic applications used in preparing nucleic acids for subsequent hybridization or cloning experiments. It a useful compound for density gradient centrifugation, for fractionation of ribosomal subunits, proteins, glycoproteins, and viruses, evacuation of radio tubes, in the preparation of electrically conducting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1303°C
Melting Point 646°C
Physical Form Beads
Quantity 5 g
Merck Index 14,2011
Solubility Information Soluble in water.
Formula Weight 168.36
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Refractive Index 1.6418
Sensitivity Hygroscopic
Density 3.99 g/mL
Assay (metals basis)
Chemical Name or Material Cesium chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'