missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cesium bromide, 99.999% (metals basis)

Brand:  Alfa Aesar™ 087639.18

Code : NEW

Additional Details : CAS Number : 7787-69-1 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11305009

  • £288.00 / 50g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

Cesium bromide
BrCs
212.81
14,2009
7787-69-1
CsBr
MFCD00010954

Specifications

Cesium bromide
99.999%
BrCs
212.81
14,2009
−4 Mesh
1.6984
1300°C
636°C
7787-69-1
(metals basis)
CsBr
MFCD00010954
4.44g/mL
Odorless
Hygroscopic
White
50g
Videos
SDS