missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium acetate, 99.998% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Cesium acetate is used in organic synthesis especially in the Perkin reaction to prepare unsaturated cinnamic-type acids. It is used in petroleum drilling fluids as an alternative to cesium formate. In organometallic chemistry, it is an additive for metal-ctalyzed coupling reactions. It is also used in the inversion of stereo chemistry of secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 194°C |
Physical Form | Lump/Powder |
Quantity | 25 g |
Solubility Information | Soluble in water and dimethyl sulfoxide. |
Formula Weight | 191.95 |
Percent Purity | 99.998% |
Odor | Weak |
Assay | (metals basis) |
Chemical Name or Material | Cesium acetate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'