missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ceruloplasmin Colorimetric Activity Kit

Assay Ceruloplasmin Activity in Serum and Urine samples.

Brand:  Invitrogen™ EIACPLC

Additional Details : Weight : 0.33820kg

Product Code. 15890893

  • £592.00 / Pack of 2
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Includes: 96 Well Plates, standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Description

Description

  • Assay Type: Enzyme Activity kit
  • Sample Types: Serum and Urine samples
  • Sensitivity: 3.26 mU/mL
  • Standard Curve Range: 31.2 mU/mL–1,000 mU/mL
Specifications

Specifications

96-well plate(s), standard, detection reagent, assay buffer, substrate, and lot-specific technical data sheet.

Store at –20°C.
Assay Activity Kit
Other Biological Fluids, Plasma, Serum
Approved for shipment on Wet or Dry Ice
Colorimetric
Ceruloplasmin Colorimetric Activity Kit
Microplate Reader
Ceruloplasmin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.