Learn More
Alfa Aesar™ Cerium vanadium oxide, 99.9% (REO)
Brand: Alfa Aesar™ 040209.14
Code : NEW
Additional Details : CAS Number : 13597-19-8 Weight : 0.02500kg Transport : UN number : 3285 Chem class : 1 Pack group : III
Quantity | 25g |
---|
Chemical Identifiers
Cerium vanadium oxide | |
255.06 | |
UN3285 |
13597-19-8 | |
MFCD00171262 | |
Cerium vanadate |
Specifications
Cerium vanadium oxide | |
Powder | |
(REO) | |
MFCD00171262 | |
Cerium vanadate | |
25g |
13597-19-8 | |
99.9% | |
255.06 | |
UN3285 | |
−200 Mesh |