missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cerium vanadium oxide, 99.9% (REO)

Brand:  Alfa Aesar™ 040209.22

Code : NEW

Additional Details : CAS Number : 13597-19-8 Weight : 0.10000kg Transport : UN number : 3285 Chem class : 1 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11339357

  • £218.36 / 100g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Cerium vanadium oxide
255.06
UN3285
13597-19-8
MFCD00171262
Cerium vanadate

Specifications

Cerium vanadium oxide
Powder
(REO)
MFCD00171262
Cerium vanadate
100g
13597-19-8
99.9%
255.06
UN3285
−200 Mesh
Videos
SDS